Methyl docosapentaenoate (7c,10c,13c,16c,19c) ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Methyl(7Z,10Z,13Z,16Z,19Z)-docosa-7,10,13,16,19-pentaenoate, all-cis-7,10,13,16,19-Methyl docosapentaenoate

202205100MG
100274-900EA 604.15 USD
100274-900
Methyl docosapentaenoate (7c,10c,13c,16c,19c) ≥99%
Methyl docosapentaenoate (7c,10c,13c,16c,19c)
MDL Number: MFCD01310506
CAS Number: 108698-02-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab964bc814430ac44b40385e395f95738b9f3347&vid=ab964bc814430ac44b40385e395f95738b9f3347&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR