2-Chloro-5-(trifluoromethyl)benzoic acid ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 2-Chloro-5-trifluoromethylbenzoic acid, 6-Chloro-α,α,α-trifluoro-m-toluic acid

0932735GM
100274-610EA 111.15 USD
100274-610
2-Chloro-5-(trifluoromethyl)benzoic acid ≥98%
2-Chloro-5-(trifluoromethyl)benzoic acid
Formula: C₈H₄ClF₃O₂
Melting Pt: 93…96 °C
MDL Number: MFCD00061094
CAS Number: 657-06-7
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=703b578f76a30f4fd6c56525049fc6d7687d7fb7&vid=703b578f76a30f4fd6c56525049fc6d7687d7fb7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR