N-Methyl-2-(trifluoromethoxy)aniline ≥97%

Supplier: INDOFINE CHEMICAL COMPANY MS

0916761GM
100274-490EA 275.92 USD
100274-490
N-Methyl-2-(trifluoromethoxy)aniline ≥97%
N-Methyl-2-(trifluoromethoxy)aniline
Formula: C₈H₈F₃NO
MDL Number: MFCD00052334
CAS Number: 175278-04-3
UN: 12352100
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=55b376e810bc291d05953bef4d81ce0d82a2df02&vid=55b376e810bc291d05953bef4d81ce0d82a2df02&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR