2,6,10,14-Tetramethylpentadecane ≥98%, liquid

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Pristane

051504100MG
100274-328EA 197.46 USD
100274-328
2,6,10,14-Tetramethylpentadecane ≥98%, liquid
2,6,10,14-Tetramethylpentadecane

  • Liquid
  • Store at +4 °C

Formula: C₁₉H₄₀
MW: 268.53 g/mol
Boiling Pt: 296
MDL Number: MFCD00008952
CAS Number: 1921-70-6
UN: 12352100
Merck Index: 12,07932
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dff1dfb3d6536cd7ab6765a338fa4f91ba7f0246&vid=dff1dfb3d6536cd7ab6765a338fa4f91ba7f0246&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR