n-Decane ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

051000100MG
100274-322EA 115.08 USD
100274-322
n-Decane ≥99%
n-Decane
Formula: H₃C(CH₂)₈CH₃
MDL Number: MFCD00008954
CAS Number: 124-18-5
UN: 2247
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5a09808f9601582d5ce92c6eabee8501d76c0557&vid=5a09808f9601582d5ce92c6eabee8501d76c0557&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR