n-Octane ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

050800100MG
100274-320EA 111.15 USD
100274-320
n-Octane ≥99%
n-Octane

Storage conditions: Store in a cool, dry place

Formula: H₃C(CH₂)₆CH₃
MW: 114.23 g/mol
MDL Number: MFCD00009556
CAS Number: 111-65-9
UN: 1262
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=253a417f96a519398f1a9061aabca3fa99282372&vid=253a417f96a519398f1a9061aabca3fa99282372&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR