Nɑ-(tert-Butoxycarbonyl)-Ndelta-carbobenzoxy-L-ornithine ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Boc-Orn(Z)-OH, Nɑ-Boc-Nδ-Cbz-L-ornithine, Nδ-Benzyloxycarbonyl-Nα-Boc-L-ornithine

048285GM
100274-290EA 111.15 USD
100274-290
Nɑ-(tert-Butoxycarbonyl)-Ndelta-carbobenzoxy-L-ornithine ≥99%
Nalpha-(tert-Butoxycarbonyl)-Ndelta-carbobenzoxy-L-ornithine
Formula: C₁₈H₂₆N₂O₆
MW: 366.41 g/mol
MDL Number: MFCD00038259
CAS Number: 2480-93-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR