N-(tert-Butoxycarbonyl)-4-fluoro-D-phenylalanine ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Boc-D-Phe(4-F)-OH, N-Boc-4-fluoro-D-phenylalanine

046601GM
100274-262EA 116.38 USD
100274-262
N-(tert-Butoxycarbonyl)-4-fluoro-D-phenylalanine ≥99%
N-(tert-Butoxycarbonyl)-4-fluoro-D-phenylalanine
Formula: C₁₄H₁₈FNO₄
MW: 283.30 g/mol
MDL Number: MFCD00079669
CAS Number: 57292-45-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f0ce07f018e6f48db42753c9a6c71f35511330a&vid=2f0ce07f018e6f48db42753c9a6c71f35511330a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR