Z-L-Lys(Tfa)-OH ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Z-Lys(Tfa)-OH

0420295GM
100273-978EA 243.23 USD
100273-978
Z-L-Lys(Tfa)-OH ≥99%
Z-L-Lys(Tfa)-OH
Formula: C₁₆H₁₉N₂O₅F₃
MW: 376.31 g/mol
MDL Number: MFCD00270524
CAS Number: 14905-30-7
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e5bd2a38a16e9decbd50cb4b4b5a110bec94dfc6&vid=6ce4905e7ec09ebb08d684fd11371beda9dc67a9&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=693de70bc04e1b59f498ec5752978bc29b90609b&vid=23db541dfc24d023b4e47d5efa1c74182b3c902c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR