N-Fmoc-D-glutamic acid 5-tert-butyl ester ≥99%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 5-tert-Butyl N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-glutamate, 5-tert-Butyl N-Fmoc-D-glutamate, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-glutamic Acid 5-tert-Butyl Ester, Fmoc-D-Glu(OtBu)-OH, (R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-(tert-butoxy)-5-oxopentanoic acid

0412711GM
100273-808EA 96.77 USD
100273-808
N-Fmoc-D-glutamic acid 5-tert-butyl ester ≥99%, powder
N-Fmoc-D-glutamic acid 5-tert-butyl ester

  • White powder
  • Store at 0-8 °C

Formula: C₂₄H₂₇NO₆
MW: 425.48 g/mol
Melting Pt: 85…90 °C
Storage Temperature: Ambient
MDL Number: MFCD00077055
CAS Number: 104091-08-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b161c29db86ef316e3e000e0afc1c138960b329f&vid=b161c29db86ef316e3e000e0afc1c138960b329f&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e2a8e2df8a7c9d03f9731010706d745922c7052e&vid=e2a8e2df8a7c9d03f9731010706d745922c7052e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR