Methyl-11-cis-octadecenoate ≥99%, liquid

Supplier: INDOFINE CHEMICAL COMPANY MS

201812100MG
100273-378EA 349.15 USD
100273-378
Methyl-11-cis-octadecenoate ≥99%, liquid
Methyl-11-cis-octadecenoate

  • "ERROR:#N/A"

Formula: C₁₉H₃₆O₂
Storage Temperature: Freezer
CAS Number: 1737-63-9
UN: 12352100
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR