trans-Vaccenic acid ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: trans-Octadec-11-enoic acid, Trans-11-Octadecenoic acid

101812100MG
100273-368EA 320.38 USD
100273-368
trans-Vaccenic acid ≥99%
trans-Vaccenic acid

Store in a cool, dry place

Formula: C₁₈H₃₄O₂
MW: 282.468 g/mol
MDL Number: MFCD00002734
CAS Number: 693-72-1
UN: 12352100

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR