3,4,5-Trifluoroanisole

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 1,2,3-Trifluoro-5-methoxybenzene, Trifluoro-3,4,5-anisole, 1-Methoxy-3,4,5-trifluorobenzene

075365GM
100273-056EA 116.38 USD
100273-056
3,4,5-Trifluoroanisole
3,4,5-Trifluoroanisole
Formula: C₇H₅F₃O
MW: 162.11
MDL Number: MFCD00042397
CAS Number: 203245-17-4
UN: 3271
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4256f94e81fb080f27e0112b8b35ee79e9194ece&vid=4256f94e81fb080f27e0112b8b35ee79e9194ece&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR