Z-Thr(tBu)-OH·DCHA ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: N-Benzyloxycarbonyl-O-tert-butyl-L-threonine dicyclohexylammonium salt, Z-Thr(tBu)-OH·DCHA, Dicyclohexylamine (2S,3R)-2-(((benzyloxy)carbonyl)amino)-3-(tert-butoxy)butanoate

049795GM
100273-008EA 109.85 USD
100273-008
Z-Thr(tBu)-OH·DCHA ≥99%
Z-Thr(tBu)-OH·DCHA
Formula: C₂₈H₄₆N₂O₅
MW: 490.685 g/mol
MDL Number: MFCD00077043
CAS Number: 16966-07-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR