Z-D-Lys(Boc)-OH ≥99% (by HPLC), powder

Supplier: INDOFINE CHEMICAL COMPANY MS

047671GM
100272-986EA 198.77 USD
100272-986
Z-D-Lys(Boc)-OH ≥99% (by HPLC), powder
Z-D-Lys(Boc)-OH

  • White powder
  • Store at 0-8 °C

Formula: C₁₉H₂₈N₂O₆
Melting Pt: 59…68 °C
MDL Number: MFCD00065699
CAS Number: 66845-42-9
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=52e55585f8537b343f85d01bfdee73ad162711a7&vid=3309e036a65f9ea7f0f2e90e4af4ec4b10afd741&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR