Z-Tyr(tBu)-OMe ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: N-Benzyloxycarbonyl-O-tert-butyl-L-tyrosine methyl ester, (S)-Methyl 2-(((benzyloxy)carbonyl)amino)-3-(4-(tert-butoxy)phenyl)propanoate

0426435GM
100272-922EA 232.77 USD
100272-922
Z-Tyr(tBu)-OMe ≥99%
Z-Tyr(tBu)-OMe
Formula: C₂₂H₂₇NO₅
MW: 385.46 g/mol
MDL Number: MFCD00237370
CAS Number: 5068-29-1
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5254024f4366b64a1a14657086d0f03ef7eccdd5&vid=297991f0edbd087661c8b95f30157c0c8a759bde&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR