L-Chg-OMe·HCI ≥99%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: H-Chg-OMe·HCl, H-Chg-OMe.HCl

0410701GM
100272-576EA 85 USD
100272-576
L-Chg-OMe·HCI ≥99%, powder
L-Chg-OMe·HCI

  • White powder
  • Store at 0-8 °C

Formula: C₉H₁₇NO₂·HCl
MW: 207.74 g/mol
Melting Pt: 185…191 °C
MDL Number: MFCD00797545
CAS Number: 14328-63-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c74945c093ea11f73d14324c7171d2697f419b16&vid=c74945c093ea11f73d14324c7171d2697f419b16&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR