Plant sterol mixture

Supplier: INDOFINE CHEMICAL COMPANY MS

60100325MG
100272-526EA 699.62 USD
100272-526
Plant sterol mixture
Plant sterol mixture

Store in a cool, dry place

MDL Number: MFCD00677687
UN: 12352100
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR