2-(3-Trifluoromethylphenyl)ethylamine ≥97%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 3-(Trifluoromethyl)phenethylamine

2619401GM
100272-252EA 158.23 USD
100272-252
2-(3-Trifluoromethylphenyl)ethylamine ≥97%
2-(3-Trifluoromethylphenyl)ethylamine
Formula: C₉H₁₀F₃N
MW: 189.18 g/mol
MDL Number: MFCD00040756
CAS Number: 52516-30-0
UN: 12352100

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR