4'-(2-Fluorophenyl)propiophenone ≥97%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 1-(2'-Fluorobiphenyl-4-yl)propan-1-one

0935441GM
100271-972EA 109.85 USD
100271-972
4'-(2-Fluorophenyl)propiophenone ≥97%
4'-(2-Fluorophenyl)propiophenone
Formula: C₁₅H₁₃FO
Melting Pt: 76…77 °C
MDL Number: MFCD00236612
CAS Number: 37989-92-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cae7e7342abafc4cc882ef0ddecb8612413a5714&vid=cae7e7342abafc4cc882ef0ddecb8612413a5714&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=907a9d8448835e47bd7e2f95758ac9e98aedf0c9&vid=907a9d8448835e47bd7e2f95758ac9e98aedf0c9&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR