4'-Iodo-2'-(trifluoromethyl)acetanilide ≥97%

Supplier: INDOFINE CHEMICAL COMPANY MS

0915921GM
100271-808EA 122.92 USD
100271-808
4'-Iodo-2'-(trifluoromethyl)acetanilide ≥97%
4'-Iodo-2'-(trifluoromethyl)acetanilide
Formula: C₉H₇F₃INO
Melting Pt: 134…136 °C
MDL Number: MFCD00052845
CAS Number: 97760-98-0
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5eedea7faba16a172f89fcefc5f366f28a0bbe67&vid=5eedea7faba16a172f89fcefc5f366f28a0bbe67&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=993c84b751e29be8f2ad2c2b01339cd2b417966e&vid=993c84b751e29be8f2ad2c2b01339cd2b417966e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR