Z-D-Ser(tBu)-OH ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Cbz-D-Ser(TBu)-OH, Z-D-Ser(TBu)-OH, Nα-Benzyloxycarbonyl-O-tert-butyl-D-serine

049671GM
100271-692EA 138.62 USD
100271-692
Z-D-Ser(tBu)-OH ≥99%
Z-D-Ser(tBu)-OH
Formula: C₁₅H₂₁NO₅
MW: 295.335 g/mol
MDL Number: MFCD00077038
CAS Number: 65806-90-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c497301b31b594f2ec5ea20b143238c542285073&vid=e6cfec9914032da601f1775ffae90cf59245a7c3&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f45f5d1bba933c3e041b9582ef0aae8b75914e4&vid=d928ee5d7e8af0634ee96ff697dfa5043d88fb04&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR