(S)-Benzyl-2-carbamoylpyrrolidine-1-carboxylate ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

0423605GM
100271-408EA 138.62 USD
100271-408
(S)-Benzyl-2-carbamoylpyrrolidine-1-carboxylate ≥99%
(S)-Benzyl-2-carbamoylpyrrolidine-1-carboxylate
Formula: C₁₃H₁₆N₂O₃
MDL Number: MFCD00020830
CAS Number: 34079-31-7
UN: 12352100
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d0c8d3dbf809671ae561ea48f42d44fd6daf7184&vid=df0f417fd1a7fa267263fa86caf5d9ed33f5712d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR