4-Nitro-DL-phenylalanine ≥99% (by HPLC), powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: H-DL-Phe(4-NO₂)-OH, H-p-Nitro-DL-Phe-OH, H-DL-Phe(4-NO2)-OH

0421775GM
100271-340EA 102 USD
100271-340
4-Nitro-DL-phenylalanine ≥99% (by HPLC), powder
4-Nitro-DL-phenylalanine

  • Faint yellow powder
  • Store at 0-8 °C

Formula: C₉H₁₀N₂O₄
MW: 210.189 g/mol
Melting Pt: 232…237 °C
MDL Number: MFCD00007384
CAS Number: 2922-40-9
UN: 12352100
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0ee84a92d53cf9707876c2f32589c198e3a8d30f&vid=8f7dc64a6c00cb0b8e4a83dc449d6420d9715b00&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR