N(α)-Benzyloxycarbonyl-D-lysine ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Cbz-D-Lys-OH, Z-D-Lys-OH

0420251GM
100271-274EA 141.23 USD
100271-274
N(α)-Benzyloxycarbonyl-D-lysine ≥99%
N(α)-Benzyloxycarbonyl-D-lysine
Formula: C₁₄H₂₀N₂O₄
MW: 280.3 g/mol
MDL Number: MFCD00067713
CAS Number: 70671-54-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR