n-Hentriacontane ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

05310025MG
100270-576EA 451.15 USD
100270-576
n-Hentriacontane ≥98%
n-Hentriacontane

Store in a cool, dry place

Formula: H₃C(CH₂)₂₉CH₃
MW: 436.85 g/mol
MDL Number: MFCD00048745
CAS Number: 630-04-6
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fcfa04487c80e92a0ab8003ccb179cfb6a0d4743&vid=fcfa04487c80e92a0ab8003ccb179cfb6a0d4743&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR