2'-Hydroxy-α-naphthoflavone ≥98%, crystals

Supplier: INDOFINE CHEMICAL COMPANY MS

H510100MG
100270-422EA 54.92 USD
100270-422
2'-Hydroxy-α-naphthoflavone ≥98%, crystals
2'-Hydroxy-alpha-naphthoflavone

White crystals


Store in a cool, dry place

Formula: C₁₉H₁₂O₃
MW: 288.29 g/mol
Melting Pt: 299…300 °C
MDL Number: MFCD00270172
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=09585b8eea27b6eb6785749b6ae4131833005a85&vid=6c21a29244735b579718a3e3f945058edfb0afb4&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR