Vanillin ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 4-Hydroxy-3-methoxybenzaldehyde

026179100GM
100270-080EA 137.31 USD
100270-080
Vanillin ≥99%
Vanillin
Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 285
Melting Pt: 82
Flash Pt: 153
MDL Number: MFCD00006942
CAS Number: 121-33-5
UN: 12352100
Merck Index: 13,09998
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd6583c5aea50cd090e103650a05eab491b4d38e&vid=535f9a4c80d51d7343126ae5e79c6c83b3beefa7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR