2,6-Dihydroxybenzoic acid ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

0260495GM
100270-066EA 45.77 USD
100270-066
2,6-Dihydroxybenzoic acid ≥98%
2,6-Dihydroxybenzoic acid
Formula: C₇H₆O₄
MW: 154.12 g/mol
Boiling Pt: 339
MDL Number: MFCD00002462
CAS Number: 303-07-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7647cfcb37983f57d88db96c7c06d97d6439369&vid=a7647cfcb37983f57d88db96c7c06d97d6439369&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR