EZ-Rate™ Westergren Pipettes, Globe Scientific
Supplier: Globe Scientific
3480
10027-470BX 219.96 USD
10027-470
EZ-Rate™ Westergren Pipettes, Globe Scientific
Pipettes Haemometer Pipettes
The EZ-Rate™ offers a simple, safe, economical and highly-accurate method for performing Westergren ESR determinations directly in 13 mm sodium citrate blood collection tubes.
The polystyrene EZ-Rate™ pipette is graduated from 0 to 180 mm and features a unique fibrous plug located at the zero mark. The plug is specifically designed to be a self-sealing aerosol barrier that stops hazardous substances from escaping through the top of the pipette.