α-Terpinene ≥90%, liquid

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: p-Mentha-1,3-diene, 1-Isopropyl-4-methylcyclohexa-1,3-diene

025064S1GM
100269-968EA 98.08 USD
100269-968
α-Terpinene ≥90%, liquid
α-Terpinene

  • Liquid
  • Store at 2-8 °C

Formula: C₁₀H₁₆
MW: 136.24 g/mol
MDL Number: MFCD00001534
CAS Number: 99-86-5
UN: 2319
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d060c0976450f111984eca48792b37e9324584d&vid=6d060c0976450f111984eca48792b37e9324584d&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR