5-Methoxypiperonal ≥97%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 7-Methoxybenzo[d][1,3]dioxole-5-carbaldehyde

0250481GM
100269-958EA 167.38 USD
100269-958
5-Methoxypiperonal ≥97%
5-Methoxypiperonal
Formula: C₉H₈O₄
MW: 180.16 g/mol
Melting Pt: 129…132 °C
MDL Number: MFCD00016900
CAS Number: 5780-07-4
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a63dee793cf2bdf69897489370565d53c74db334&vid=a63dee793cf2bdf69897489370565d53c74db334&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR