Cholesterol acetate ≥97%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: (3S,8S,9S,10R,13R,14S,17R)-10,13-Dimethyl-17-((R)-6-methylheptan-2-yl)-2,3,4,7,8,9,10,11,12,13,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-3-yl acetate, Cholesteryl Acetate, Acetyl Cholesterol, Acetic Acid Cholesterol Ester

02356225GM
100269-878EA 338.69 USD
100269-878
Cholesterol acetate ≥97%
Cholesterol acetate
Formula: C₂₉H₄₈O₂
MW: 428.70 g/mol
Melting Pt: 112…114 °C
MDL Number: MFCD00003636
CAS Number: 604-35-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=81ea5b2f21ad1fd347ba9fe976c946ae13816259&vid=81ea5b2f21ad1fd347ba9fe976c946ae13816259&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR