(+)-Isocorydine hydrochloride ≥95%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

02185310MG
100269-820EA 666.92 USD
100269-820
(+)-Isocorydine hydrochloride ≥95%, powder
(+)-Isocorydine hydrochloride

  • Powder
  • Store in a cool, dry place

Formula: C₂₀H₂₃NO₄·HCl
MDL Number: MFCD00134198
CAS Number: 13552-72-2
UN: 12352100
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fb109b1fd943a4aae6ac53602b6c8243b6d6a5c7&vid=fb109b1fd943a4aae6ac53602b6c8243b6d6a5c7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR