Senecionine ≥95%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

02182810MG
100269-818EA 634.23 USD
100269-818
Senecionine ≥95%, powder
Senecionine

  • Powder
  • Store in a cool, dry place

Formula: C₁₈H₂₅NO₅
Melting Pt: 228…236 °C
MDL Number: MFCD00221720
CAS Number: 130-01-8
UN: 12352100
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR