Cytochalasin D ≥98%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 7(S),18(R)-Dihydroxy-16(S),18-dimethyl-10-phenyl[11]cytochalasa-6(12),13(E),19(E)-triene-1,17-dione 21(R)-acetate

0215641MG
100269-800EA 452.46 USD
100269-800
Cytochalasin D ≥98%, powder
Cytochalasin D

  • Powder
  • Store at -20°C

Formula: C₃₀H₃₇NO₆
MW: 507.63
Melting Pt: 255…260 °C
Storage Temperature: Freezer
MDL Number: MFCD00077706
CAS Number: 22144-77-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e61243c3859df2e28533c9707741b37af283f63f&vid=e61243c3859df2e28533c9707741b37af283f63f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR