(±)-Naringenin ≥99% (by HPLC), crystalline powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: (±)-2,3-Dihydro-5,7-dihydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, 5,7-Dihydroxy-2-(4-hydroxyphenyl)chroman-4-one, 4',5,7-Trihydroxyflavanone, Naringenin

021128S10MG
100269-782EA 137.31 USD
100269-782
(±)-Naringenin ≥99% (by HPLC), crystalline powder
(±)-Naringenin

  • Brown crystalline powder
  • Store in a cool, dry place

Formula: C₁₅H₁₂O₅
MW: 272.25 g/mol
Melting Pt: 254…256 °C
MDL Number: MFCD00006844
CAS Number: 67604-48-2
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b84004e39e16e21d54c5c9d0d1a349ab068380e9&vid=b84004e39e16e21d54c5c9d0d1a349ab068380e9&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR