Tiliroside ≥99% (by HPLC), powder

Supplier: INDOFINE CHEMICAL COMPANY MS

021001S10MG
100269-768EA 402.77 USD
100269-768
Tiliroside ≥99% (by HPLC), powder
Tiliroside

  • Yellow powder
  • Store in a cool, dry place

Formula: C₃₀H₂₆O₁₃
Melting Pt: 257…260 °C
MDL Number: MFCD00017454
CAS Number: 20316-62-5
UN: 12352100
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a85b62223f9d12755fa878897db67de52327a291&vid=a85b62223f9d12755fa878897db67de52327a291&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd6cedd37079b7d52f3bc3fbb03a29d7b38c79f9&vid=dd6cedd37079b7d52f3bc3fbb03a29d7b38c79f9&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR