Cyanidin-3-O-galactoside chloride ≥97%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 2-(3,4-Dihydroxyphenyl)-5,7-dihydroxy-3-(((2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)chromenylium chloride

020923S5MG
100269-758EA 490.38 USD
100269-758
Cyanidin-3-O-galactoside chloride ≥97%, powder
Cyanidin-3-O-galactoside chloride

  • Black red powder
  • Store at -20 °C, To be kept away from light and moisture

Formula: C₂₁H₂₁O₁₁Cl
MW: 484.85 g/mol
MDL Number: MFCD00270387
CAS Number: 27661-36-5
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=588fcba82dfc995bf05042c370738070bb62d98c&vid=588fcba82dfc995bf05042c370738070bb62d98c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR