3-Bromo-4-ethoxybenzoic acid ≥99%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

195841GM
100269-466EA 36.62 USD
100269-466
3-Bromo-4-ethoxybenzoic acid ≥99%, powder
3-Bromo-4-ethoxybenzoic acid

White powder


Store in a cool, dry place

Formula: C₉H₉BrO₃
MW: 245.08 g/mol
Melting Pt: 213.8…215.2 °C
MDL Number: MFCD01143606
CAS Number: 24507-29-7
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=82ddb3e70ceb119f872a7957c087111e75c1639f&vid=82ddb3e70ceb119f872a7957c087111e75c1639f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR