3-Bromo-4',6'-Dimethoxy-2'-Hydroxychalcone ≥98%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

19485100MG
100269-418EA 43.15 USD
100269-418
3-Bromo-4',6'-Dimethoxy-2'-Hydroxychalcone ≥98%, powder
3-Bromo-4',6'-Dimethoxy-2'-Hydroxychalcone

Pale yellow powder


Store in a cool, dry place

Formula: C₁₇H₁₅BrO₄
Melting Pt: 108…110 °C
MDL Number: MFCD03788885
UN: 12352100
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f3c850b1288034e00dd84455060401bd89c6b646&vid=a716ee68ccaa78e949ce0803f6eaa5b37878c18d&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR