5-Bromo-2,2'-dihydroxy-4',6'-dimethoxychalcone ≥98%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

19484100MG
100269-414EA 47.08 USD
100269-414
5-Bromo-2,2'-dihydroxy-4',6'-dimethoxychalcone ≥98%, powder
5-Bromo-2,2'-dihydroxy-4',6'-dimethoxychalcone

Yellow powder


Store in a cool, dry place

Formula: C₁₇H₁₅BrO₅
MW: 379.21 g/mol
Melting Pt: 170…173 °C
MDL Number: MFCD03788884
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8e61308721740fd616cf84f1f6288d3f4a20389c&vid=9f56a49f5d785dfc88265e74e65cbb7f01b27b94&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR