3(4'-Bromophenyl)-6-ethoxy-4-methylcoumarin ≥98%, crystalline powder

Supplier: INDOFINE CHEMICAL COMPANY MS

19241100MG
100269-288EA 107.23 USD
100269-288
3(4'-Bromophenyl)-6-ethoxy-4-methylcoumarin ≥98%, crystalline powder
3(4'-Bromophenyl)-6-ethoxy-4-methylcoumarin

Light yellow crystalline powder


Store in a cool, dry place

Formula: C₁₈H₁₅BrO₃
MW: 359.22 g/mol
Melting Pt: 161…162.5 °C
MDL Number: MFCD02660619
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e72116694efc485fa05685184e7ea5f048d6b8ad&vid=a1698d2991aec3b5085d73d8b1371e344882197d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR