3-Bromo-4-hydroxybenzoic acid ≥98%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

191555GM
100269-216EA 36.62 USD
100269-216
3-Bromo-4-hydroxybenzoic acid ≥98%, powder
3-Bromo-4-hydroxybenzoic acid

  • White powder
  • Store in a cool, dry place

Formula: C₇H₅BrO₃
MW: 217.02
Melting Pt: 177…179 °C
MDL Number: MFCD00017547
CAS Number: 14348-41-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2356bb143a82bd3ecedb17aba65d18cf97c3b4f&vid=b2356bb143a82bd3ecedb17aba65d18cf97c3b4f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR