Capsaicin ≥95% (by HPLC)

Supplier: INDOFINE CHEMICAL COMPANY MS

02898610MG
100269-068EA 119.23 USD
100269-068
Capsaicin ≥95% (by HPLC)
Capsaicin
Formula: C₁₈H₂₇NO₃
MDL Number: MFCD00017259
CAS Number: 404-86-4
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR