Methyl-3,4,5-trihydroxybenzoate ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Methyl gallate, Gallic acid methyl ester, Methyl 3,4,5-trihydroxybenzoate

026081100GM
100268-982EA 124.23 USD
100268-982
Methyl-3,4,5-trihydroxybenzoate ≥98%
Methyl 3,4,5-trihydroxybenzoate
Formula: C₈H₈O₅
MW: 184.15 g/mol
Boiling Pt: 450
Melting Pt: 198…203 °C
MDL Number: MFCD00002194
CAS Number: 99-24-1
UN: 12352100
Merck Index: 13,04366
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=94bc83df2cb039cd510e2f477897ac53da352b96&vid=94bc83df2cb039cd510e2f477897ac53da352b96&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR