(R)-(-)-Linalool ≥95%, liquid

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: (-)-Linalool, (R)-3,7-Dimethylocta-1,6-dien-3-ol, (R)-(-)-3,7-Dimethyl-1,6-octadien-3-ol, L-Linalool

0254301GM
100268-930EA 223.62 USD
100268-930
(R)-(-)-Linalool ≥95%, liquid
(R)-(-)-Linalool

  • Liquid
  • Store at 2-8 °C

Formula: C₁₀H₁₈O
MW: 154.25
MDL Number: MFCD00135469
CAS Number: 126-91-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR