N,N'-Diacetylchitobiose ≥96%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: GlcNAc beta(1-4)GlcNAc, 2-Acetamido-2-deoxy-4-O-(2-acetamido-2-deoxy-beta-D-glucopyranosyl)-D-glucopyranose

02440010MG
100268-834EA 997.77 USD
100268-834
N,N'-Diacetylchitobiose ≥96%
N,N'-Diacetylchitobiose
Formula: C₁₆H₂₈N₂O₁₁
MW: 424.40 g/mol
MDL Number: MFCD00077715
CAS Number: 35061-50-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=432bc436515ab66022f4bfb099feca8e1a10a4a6&vid=432bc436515ab66022f4bfb099feca8e1a10a4a6&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR