Xylitol ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: (2R,3r,4S)-Pentane-1,2,3,4,5-pentaol

024311100GM
100268-822EA 137.31 USD
100268-822
Xylitol ≥99%
Xylitol
Formula: C₅H₁₂O₅
MW: 152.15 g/mol
Boiling Pt: 495
Melting Pt: 94
MDL Number: MFCD00064292
CAS Number: 87-99-0
UN: 12352100
Merck Index: 13,10140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=00fcdff4a89e6d5ea846d02afd596df28d4dfefb&vid=00fcdff4a89e6d5ea846d02afd596df28d4dfefb&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR