Stevioside, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: (2S,3R,4S,5S,6R)-3,4,5-Trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl (4R,4aS,6aR,9S,11aR,11bS)-9-(((2S,3R,4S,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-3-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-yl)oxy)-4,11b-dimethyl-8-methylenetetradecahydro-6a,9-methanocyclohepta[a]naphthalene-4-carboxylate

02371810MG
100268-756EA 231.46 USD
100268-756
Stevioside, powder
Stevioside

  • White fluffy powder
  • Store at 2-8 °C

Formula: C₃₈H₆₀O₁₈
MW: 804.88 g/mol
Melting Pt: 199…201 °C
MDL Number: MFCD00079561
CAS Number: 57817-89-7
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=93ec378332d70aad4ead6417747445bda266895d&vid=2d6e186e6a176c79cd2f515cd8e1d08c617ff1d9&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a365b6227607bac60dce5062b240c5a61076e9fe&vid=4cb70f0e81a7ecc9f488f5bcd7776d9f375e35a0&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR